Learn More
Medchemexpress LLC Polyoxyethylene (10) lauryl ether | 9002-92-0 | 98.0% | 25 G
SDP
Catalog No. 5000414367
Encompass_Preferred
Polyoxyethylene (10) lauryl ether is a nonionic surfactant with unique properties. It is known for dissolving liposomes and minimal interference with endotoxin detection. This product forms hybrid liposomes with Dimyristoylphosphatidylcholine (DMPC), which can inhibit the proliferation of human promyelocytic leukemia cells without the administration of any drugs.
  • Exhibits unique properties in dissolving liposomes.
  • Exerts minimal interference with endotoxin detection.
  • Forms hybrid liposomes to inhibit proliferation of human promyelocytic leukemia cells.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Recommended storage temperature: 4°C, sealed storage, away from moisture.
  • When in solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Stable under recommended storage conditions.
Catalog No. 50-004-14367 Supplier Medchemexpress LLC Supplier No. 25GHYB2106D
Polyoxyethylene (10) lauryl ether is a nonionic surfactant with unique properties. It is known for dissolving liposomes and minimal interference with endotoxin detection. This product forms hybrid liposomes with Dimyristoylphosphatidylcholine (DMPC), which can inhibit the proliferation of human promyelocytic leukemia cells without the administration of any drugs.
  • Exhibits unique properties in dissolving liposomes.
  • Exerts minimal interference with endotoxin detection.
  • Forms hybrid liposomes to inhibit proliferation of human promyelocytic leukemia cells.
  • Keep container tightly sealed in cool, well-ventilated area.
  • Store away from direct sunlight and sources of ignition.
  • Recommended storage temperature: 4°C, sealed storage, away from moisture.
  • When in solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Stable under recommended storage conditions.