Learn More
Medchemexpress LLC Polyoxyethylene(10) oleyl ether | 9004-98-2 | MFCD00074369 | >99.0% | 709 (average) | (C2H4O)XC18H36O | 10 G
SDP
Catalog No. 5000282106
Encompass_Preferred
Polyoxyethylene(10) oleyl ether is a nonionic surfactant composed of an oleyl residue attached to a polyethylene glycol chain of about 10 ethylene oxide units. It is used as an emulsifier and solubilizer in laboratory formulations and research applications where improved wetting, dispersion, or solubility of hydrophobic compounds is required.
  • Nonionic surfactant with a decaethylene glycol chain.
  • Good emulsifying and solubilizing properties.
  • Useful as a wetting and dispersing agent in formulations.
  • Biodegradable and low toxicity profile for many applications.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-82106 Supplier Medchemexpress LLC Supplier No. HYW25016810G
Polyoxyethylene(10) oleyl ether is a nonionic surfactant composed of an oleyl residue attached to a polyethylene glycol chain of about 10 ethylene oxide units. It is used as an emulsifier and solubilizer in laboratory formulations and research applications where improved wetting, dispersion, or solubility of hydrophobic compounds is required.
  • Nonionic surfactant with a decaethylene glycol chain.
  • Good emulsifying and solubilizing properties.
  • Useful as a wetting and dispersing agent in formulations.
  • Biodegradable and low toxicity profile for many applications.
  • Available in small laboratory pack sizes for research use.