Learn More
Medchemexpress LLC Polyoxyethylene (100) stearyl ether | 9005-00-9 | MFCD00074177 | 98.0% | Average Mn ~4,670 g/mol | C20H42O2 | 10 G
SDP
Catalog No. 5000281607
Encompass_Preferred
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether) used as an emulsifier and solubilizer in laboratory and biochemical formulations. It is a high-molecular-weight PEG derivative with supporting safety and technical documentation available.
  • Nonionic surfactant for emulsification and solubilization in aqueous systems.
  • Average Mn ≈4,670 g/mol for consistent performance.
  • Typical purity around 98.0% as supplied for laboratory use.
  • Solid or waxy form suitable for formulation and surface-active applications.
  • Available in small-scale packages with accompanying SDS and data sheet.
Catalog No. 50-002-81607 Supplier Medchemexpress LLC Supplier No. HYW250183A10G
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether) used as an emulsifier and solubilizer in laboratory and biochemical formulations. It is a high-molecular-weight PEG derivative with supporting safety and technical documentation available.
  • Nonionic surfactant for emulsification and solubilization in aqueous systems.
  • Average Mn ≈4,670 g/mol for consistent performance.
  • Typical purity around 98.0% as supplied for laboratory use.
  • Solid or waxy form suitable for formulation and surface-active applications.
  • Available in small-scale packages with accompanying SDS and data sheet.