Learn More
Medchemexpress LLC Polyoxyethylene (100) stearyl ether | 9005-00-9 | MFCD00074177 | Average Mn ≈4,670 g/mol | (C2H4O)nC18H38O | 25 G
SDP
Catalog No. 5000281795
Encompass_Preferred
Polyoxyethylene (100) stearyl ether is a nonionic ethoxylated surfactant (average Mn ≈4,670) used as an emulsifier and formulation excipient in laboratory research, especially for liposome preparation and biochemical formulations where a high-molecular-weight polyoxyethylene stearyl ether is required.
  • Nonionic surfactant suitable for emulsification and solubilization.
  • High average molecular weight for increased steric stabilization.
  • Suitable for liposome preparation and formulation work.
  • Available in small laboratory pack sizes for research use.
  • Polymeric structure provides flexible formulation properties.
Catalog No. 50-002-81795 Supplier Medchemexpress LLC Supplier No. HYW250183A25G
Polyoxyethylene (100) stearyl ether is a nonionic ethoxylated surfactant (average Mn ≈4,670) used as an emulsifier and formulation excipient in laboratory research, especially for liposome preparation and biochemical formulations where a high-molecular-weight polyoxyethylene stearyl ether is required.
  • Nonionic surfactant suitable for emulsification and solubilization.
  • High average molecular weight for increased steric stabilization.
  • Suitable for liposome preparation and formulation work.
  • Available in small laboratory pack sizes for research use.
  • Polymeric structure provides flexible formulation properties.