Learn More
Medchemexpress LLC Polyoxyethylene(20) oleyl ether | 9004-98-2 | MFCD00074369 | 1150 (average) | C58H116O21 | 25 G
SDP
Catalog No. 5000282108
Encompass_Preferred
Polyoxyethylene(20) oleyl ether is a nonionic surfactant (average Mn ≈ 1150, CAS 9004-98-2) used in laboratory research for emulsification, solubilization, and detergency. It is supplied for research use only and is commonly used in biochemical assays and formulation development.
  • Nonionic surfactant with emulsification and detergency properties.
  • Average molecular weight approximately 1150 (n ≈ 20).
  • Used as an emulsifier, solubilizer, and detergent in formulations.
  • Intended for research use only.
  • Available in small packaging sizes suitable for laboratory use.
Catalog No. 50-002-82108 Supplier Medchemexpress LLC Supplier No. HYW250168A25G
Polyoxyethylene(20) oleyl ether is a nonionic surfactant (average Mn ≈ 1150, CAS 9004-98-2) used in laboratory research for emulsification, solubilization, and detergency. It is supplied for research use only and is commonly used in biochemical assays and formulation development.
  • Nonionic surfactant with emulsification and detergency properties.
  • Average molecular weight approximately 1150 (n ≈ 20).
  • Used as an emulsifier, solubilizer, and detergent in formulations.
  • Intended for research use only.
  • Available in small packaging sizes suitable for laboratory use.