Learn More
Medchemexpress LLC Polyoxyethylene (20) oleyl ether | 9004-98-2 | MFCD00074369 | (C2H4O)xC18H36O (n ≈ 20) | 5 G
SDP
Catalog No. 5000282105
Encompass_Preferred
Polyoxyethylene(20) oleyl ether is a nonionic ethoxylated surfactant used as an emulsifier, solubilizer, and detergent in laboratory and formulation work. It has an average molecular weight of about 1,150 g/mol and is solid below 25°C and liquid above 30°C. The reagent is supplied with analytical data for quality control and is intended for research use.
  • Acts as a nonionic emulsifier and solubilizer.
  • Improves solubility of hydrophobic compounds in aqueous media.
  • Compatible with a wide range of formulation types.
  • Gentle on biological samples and low in irritancy.
  • Supplied with analytical data for quality control.
Catalog No. 50-002-82105 Supplier Medchemexpress LLC Supplier No. HYW250168A5G
Polyoxyethylene(20) oleyl ether is a nonionic ethoxylated surfactant used as an emulsifier, solubilizer, and detergent in laboratory and formulation work. It has an average molecular weight of about 1,150 g/mol and is solid below 25°C and liquid above 30°C. The reagent is supplied with analytical data for quality control and is intended for research use.
  • Acts as a nonionic emulsifier and solubilizer.
  • Improves solubility of hydrophobic compounds in aqueous media.
  • Compatible with a wide range of formulation types.
  • Gentle on biological samples and low in irritancy.
  • Supplied with analytical data for quality control.