Learn More
Medchemexpress LLC Polyoxyethylene oleyl ether | 9004-98-2 | MFCD00074369 | 1,149.53 g/mol | (C2H4O)xC18H36O | 50 G
SDP
Catalog No. 5000282339
Encompass_Preferred
Polyoxyethylene(20) oleyl ether is a nonionic surfactant (polyethylene glycol oleyl ether) with average molecular weight ≈1,150 g/mol. It is commonly used in laboratory research for emulsification and detergency. Supplied for research use only.
  • Nonionic surfactant providing emulsification and detergency.
  • Average molecular weight ~1,150 g/mol.
  • CAS number 9004-98-2 for unambiguous identification.
  • Intended for laboratory research use only.
  • Available in small pack sizes suitable for bench-scale work.
Catalog No. 50-002-82339 Supplier Medchemexpress LLC Supplier No. HYW250168A50G
Polyoxyethylene(20) oleyl ether is a nonionic surfactant (polyethylene glycol oleyl ether) with average molecular weight ≈1,150 g/mol. It is commonly used in laboratory research for emulsification and detergency. Supplied for research use only.
  • Nonionic surfactant providing emulsification and detergency.
  • Average molecular weight ~1,150 g/mol.
  • CAS number 9004-98-2 for unambiguous identification.
  • Intended for laboratory research use only.
  • Available in small pack sizes suitable for bench-scale work.