Learn More
Medchemexpress LLC Polyoxyethylene Sorb 5kg
SDP
Catalog No. 5000206264
Encompass_Preferred
Polyoxyethylene Sorbitan Monostearate is an ester of the saturated fatty acid stearic acid (C18 0) Polyoxyethylene Sorbitan Monostearate can be used as an excipient such as surfactant emulsifier solubilizer wetting agent Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]
Catalog No. 50-002-06264 Supplier Medchemexpress LLC Supplier No. HYW1158185KG
Polyoxyethylene Sorbitan Monostearate is an ester of the saturated fatty acid stearic acid (C18 0) Polyoxyethylene Sorbitan Monostearate can be used as an excipient such as surfactant emulsifier solubilizer wetting agent Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]