Learn More
Medchemexpress LLC HY-101530 200mg Medchemexpress, Polyoxyethylene stearate CAS:9004-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101530200MG
Encompass_Preferred
Medchemexpress, HY-101530 200mg Polyoxyethylene stearate CAS:9004-99-3 Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2159