Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | 328.53 g/mol | C20H40O3 | 100 G
SDP
Catalog No. 5000248427
Encompass_Preferred
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent and surfactant used as an excipient to stabilize oil-in-water emulsions, improve solubilization of lipophilic ingredients, and serve as an ointment or suppository base. It is supplied as a white to off-white solid for research use.
  • Non-ionic emulsifier for oil-in-water formulations.
  • Improves solubilization of lipophilic compounds.
  • Suitable as an excipient in topical and suppository bases.
  • White to off-white solid with convenient handling properties.
  • Used in formulation development and research applications.
Catalog No. 50-002-48427 Supplier Medchemexpress LLC Supplier No. HY101530100G
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent and surfactant used as an excipient to stabilize oil-in-water emulsions, improve solubilization of lipophilic ingredients, and serve as an ointment or suppository base. It is supplied as a white to off-white solid for research use.
  • Non-ionic emulsifier for oil-in-water formulations.
  • Improves solubilization of lipophilic compounds.
  • Suitable as an excipient in topical and suppository bases.
  • White to off-white solid with convenient handling properties.
  • Used in formulation development and research applications.