Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | MFCD00148007 | 328.53 g/mol (monomer) | C20H40O3 | 10 G
SDP
Catalog No. 5000248283
Encompass_Preferred
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent used in laboratory and formulation research. It appears as a solid, white to off-white powder (CAS 9004-99-3) and is provided in multiple small pack sizes for research applications. Safety data, handling instructions, and batch-specific certificates of analysis are available from the manufacturer.
  • Non-ionic emulsifier for stabilizing water-oil systems.
  • Suitable for formulation development and surfactant studies.
  • Solid powder form with defined storage conditions for stability.
  • Available in multiple small pack sizes suitable for laboratory use.
  • Batch-specific COA and SDS available for quality and handling information.
Catalog No. 50-002-48283 Supplier Medchemexpress LLC Supplier No. HY10153010G
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent used in laboratory and formulation research. It appears as a solid, white to off-white powder (CAS 9004-99-3) and is provided in multiple small pack sizes for research applications. Safety data, handling instructions, and batch-specific certificates of analysis are available from the manufacturer.
  • Non-ionic emulsifier for stabilizing water-oil systems.
  • Suitable for formulation development and surfactant studies.
  • Solid powder form with defined storage conditions for stability.
  • Available in multiple small pack sizes suitable for laboratory use.
  • Batch-specific COA and SDS available for quality and handling information.