Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | MFCD00148007 | 622.91 g/mol | (C2H4O)nC18H36O2 | 50 G
SDP
Catalog No. 5000248265
Encompass_Preferred
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent and surfactant used as a pharmaceutical excipient to solubilize, emulsify, and stabilize oil-in-water formulations. It can aid in improving oral bioavailability of coadministered compounds and is used in topical and suppository bases.
  • Non-ionic emulsifier suitable for o/w systems.
  • Solubilizes lipophilic compounds and aids formulation.
  • Can improve oral bioavailability of coadministered drugs.
  • Suitable for ointment and suppository bases.
  • Available in multiple laboratory pack sizes for formulation work.
Catalog No. 50-002-48265 Supplier Medchemexpress LLC Supplier No. HY10153050G
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent and surfactant used as a pharmaceutical excipient to solubilize, emulsify, and stabilize oil-in-water formulations. It can aid in improving oral bioavailability of coadministered compounds and is used in topical and suppository bases.
  • Non-ionic emulsifier suitable for o/w systems.
  • Solubilizes lipophilic compounds and aids formulation.
  • Can improve oral bioavailability of coadministered drugs.
  • Suitable for ointment and suppository bases.
  • Available in multiple laboratory pack sizes for formulation work.