Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | MFCD00148007 | 98.0% | 328.53 g/mol | C20H40O3 | 25 G
SDP
Catalog No. 5000248975
Encompass_Preferred
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent used in pharmaceutical, cosmetic, and microbiological formulations to improve wetting and stabilize oil-in-water emulsions. It has documented uses in culture media to enhance growth of certain mycobacteria and has been studied for effects on drug efflux mechanisms, supporting formulation and delivery research.
  • Non-ionic emulsifying agent suitable for oil-in-water formulations.
  • Improves wetting and stabilizes emulsions for formulation consistency.
  • Enhances growth of certain mycobacteria in radiometric culture media.
  • Modulates P-glycoprotein (P-gp) mediated efflux, aiding delivery studies.
  • High DMSO solubility with ultrasonic aid for in vitro use.
  • Available in multiple laboratory pack sizes for research applications.
Catalog No. 50-002-48975 Supplier Medchemexpress LLC Supplier No. HY10153025G
Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent used in pharmaceutical, cosmetic, and microbiological formulations to improve wetting and stabilize oil-in-water emulsions. It has documented uses in culture media to enhance growth of certain mycobacteria and has been studied for effects on drug efflux mechanisms, supporting formulation and delivery research.
  • Non-ionic emulsifying agent suitable for oil-in-water formulations.
  • Improves wetting and stabilizes emulsions for formulation consistency.
  • Enhances growth of certain mycobacteria in radiometric culture media.
  • Modulates P-glycoprotein (P-gp) mediated efflux, aiding delivery studies.
  • High DMSO solubility with ultrasonic aid for in vitro use.
  • Available in multiple laboratory pack sizes for research applications.