Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | MFCD00148007 | 99.0% | 328.53 g/mol | C20H40O3 | 5 G
SDP
Catalog No. 5000248967
Encompass_Preferred
Polyoxyethylene stearate is a non-ionic emulsifying and solubilizing agent used as an excipient in pharmaceutical formulations and laboratory research. It facilitates oil-in-water emulsions and increases the solubility and bioavailability of lipophilic compounds in aqueous vehicles, and is supplied as a solid powder for formulation use.
  • Non-ionic emulsifier for oil-in-water formulations.
  • Solubilizer for lipophilic compounds, compatible with DMSO.
  • Can improve oral bioavailability of certain coadministered drugs.
  • Supplied as a solid powder with good handling properties.
  • Soluble in DMSO (reported up to 100 mg/mL with sonication).
  • Stable under typical refrigerated or frozen storage for extended periods.
Catalog No. 50-002-48967 Supplier Medchemexpress LLC Supplier No. HY1015305G
Polyoxyethylene stearate is a non-ionic emulsifying and solubilizing agent used as an excipient in pharmaceutical formulations and laboratory research. It facilitates oil-in-water emulsions and increases the solubility and bioavailability of lipophilic compounds in aqueous vehicles, and is supplied as a solid powder for formulation use.
  • Non-ionic emulsifier for oil-in-water formulations.
  • Solubilizer for lipophilic compounds, compatible with DMSO.
  • Can improve oral bioavailability of certain coadministered drugs.
  • Supplied as a solid powder with good handling properties.
  • Soluble in DMSO (reported up to 100 mg/mL with sonication).
  • Stable under typical refrigerated or frozen storage for extended periods.