Learn More
Medchemexpress LLC Polyoxyethylene stearyl ether | 9005-00-9 | MFCD00074177 | Average Mn ≈4670 g/mol | (C2H4O)nC18H38O | 100 G
SDP
Catalog No. 5000281815
Encompass_Preferred
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether, n≈100) used as an emulsifying and solubilizing agent in biochemical and formulation applications, including liposome preparation and other life-science research uses.
  • High average molecular weight (Mn ≈4670) suitable for stable emulsions.
  • Nonionic surfactant with low charge-related interactions.
  • Effective emulsifier and solubilizer for hydrophobic compounds.
  • Compatible with liposome formulations and biological assays.
  • Available in laboratory packaging sizes for formulation and research use.
Catalog No. 50-002-81815 Supplier Medchemexpress LLC Supplier No. HYW250183A100G
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether, n≈100) used as an emulsifying and solubilizing agent in biochemical and formulation applications, including liposome preparation and other life-science research uses.
  • High average molecular weight (Mn ≈4670) suitable for stable emulsions.
  • Nonionic surfactant with low charge-related interactions.
  • Effective emulsifier and solubilizer for hydrophobic compounds.
  • Compatible with liposome formulations and biological assays.
  • Available in laboratory packaging sizes for formulation and research use.