Learn More
Medchemexpress LLC Polyoxyethylene stearyl ether | 9005-00-9 | MFCD00074177 | Average Mn ≈711 g/mol | (C2H4O)nC18H38O | 100 G
SDP
Catalog No. 5000281673
Encompass_Preferred
Polyoxyethylene stearyl ether is a nonionic surfactant and laboratory reagent with an average Mn of about 711 g/mol. It is used in research settings for formulation, emulsification, and solubilization tasks, supplied as a 100 G package for laboratory use only.
  • Nonionic surfactant suitable for emulsification and solubilization.
  • Useful in formulation and biochemical assay preparation.
  • Low solubility in water, compatible with many organic solvents.
  • Typically supplied as a white to off-white oil for easy handling.
  • Recommended storage under inert gas and away from moisture to preserve quality.
Catalog No. 50-002-81673 Supplier Medchemexpress LLC Supplier No. HYW250183100G
Polyoxyethylene stearyl ether is a nonionic surfactant and laboratory reagent with an average Mn of about 711 g/mol. It is used in research settings for formulation, emulsification, and solubilization tasks, supplied as a 100 G package for laboratory use only.
  • Nonionic surfactant suitable for emulsification and solubilization.
  • Useful in formulation and biochemical assay preparation.
  • Low solubility in water, compatible with many organic solvents.
  • Typically supplied as a white to off-white oil for easy handling.
  • Recommended storage under inert gas and away from moisture to preserve quality.