Learn More
Medchemexpress LLC Polyoxyethylene stearyl ether | 9005-00-9 | MFCD00074177 | C18H37(OCH2CH2)nOH | 50 G
SDP
Catalog No. 5000280503
Encompass_Preferred
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether) consisting of a stearyl (C18) hydrophobic tail linked to approximately 100 ethylene oxide units (average Mn ≈4,670 g/mol). It is used as an emulsifier, solubilizer, and formulation excipient in laboratory and life-science applications. Consult the product datasheet and SDS for handling and specifications.
  • Nonionic surfactant and emulsifier.
  • Average molecular weight ≈4,670 g/mol.
  • Polyoxyethylene chain length ≈100 ethylene oxide units.
  • Useful as a solubilizer and formulation excipient.
  • Suitable for use in emulsions and nanoparticle formulations.
  • Available in 50 g pack; consult SDS for safety and handling.
Catalog No. 50-002-80503 Supplier Medchemexpress LLC Supplier No. HYW250183A50G
Polyoxyethylene (100) stearyl ether is a nonionic surfactant (polyethylene glycol octadecyl ether) consisting of a stearyl (C18) hydrophobic tail linked to approximately 100 ethylene oxide units (average Mn ≈4,670 g/mol). It is used as an emulsifier, solubilizer, and formulation excipient in laboratory and life-science applications. Consult the product datasheet and SDS for handling and specifications.
  • Nonionic surfactant and emulsifier.
  • Average molecular weight ≈4,670 g/mol.
  • Polyoxyethylene chain length ≈100 ethylene oxide units.
  • Useful as a solubilizer and formulation excipient.
  • Suitable for use in emulsions and nanoparticle formulations.
  • Available in 50 g pack; consult SDS for safety and handling.