Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polyoxyethylene tridecyl ether | 78330-21-9 | MFCD00132049 | ≥98.0% | 728.99 g/mol | C37H76O13 | 25 G
SDP

Catalog No. 5000282381
Encompass_Preferred

Polyoxyethylene tridecyl ether is a nonionic surfactant (poly(ethylene glycol) (12) tridecyl ether) used as a solubilizing and wetting agent in biochemical assays and laboratory research. It is an ethoxylated fatty alcohol supplied with batch documentation and a certificate of analysis for research use.

  • Nonionic surfactant for solubilization and wetting.
  • Suitable for biochemical assays and laboratory research.
  • High reported purity ≥98.0% (supplier COA).
  • Molecular weight 728.99 g/mol and molecular formula C37H76O13.
  • CAS number 78330-21-9 for unambiguous identification.

Catalog No. 50-002-82381 Supplier Medchemexpress LLC Supplier No. HYW25016625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polyoxyethylene tridecyl ether is a nonionic surfactant (poly(ethylene glycol) (12) tridecyl ether) used as a solubilizing and wetting agent in biochemical assays and laboratory research. It is an ethoxylated fatty alcohol supplied with batch documentation and a certificate of analysis for research use.

  • Nonionic surfactant for solubilization and wetting.
  • Suitable for biochemical assays and laboratory research.
  • High reported purity ≥98.0% (supplier COA).
  • Molecular weight 728.99 g/mol and molecular formula C37H76O13.
  • CAS number 78330-21-9 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.