Learn More
Medchemexpress LLC Polyoxyethylene tridecyl ether | 78330-21-9 | MFCD00132049 | Approximately 729 g/mol | C13H27(OCH2CH2)nOH, n≈12 | 50 G
SDP
Catalog No. 5000281302
Encompass_Preferred
Polyoxyethylene tridecyl ether is a nonionic ethoxylated fatty alcohol (commonly described as PEG-12 tridecyl ether) used as an emulsifier, solubilizer, and wetting agent in biochemical, formulation, and personal-care applications. It offers good water solubility and emulsion-stabilizing properties, and is also used as a raw material for producing other surfactants and specialty chemicals.
  • Nonionic surfactant for emulsification and solubilization.
  • Good water solubility across formulation types.
  • Low toxicity and mild handling profile.
  • Effective wetting agent for surface treatments.
  • Useful intermediate for producing specialty surfactants.
Catalog No. 50-002-81302 Supplier Medchemexpress LLC Supplier No. HYW25016650G
Polyoxyethylene tridecyl ether is a nonionic ethoxylated fatty alcohol (commonly described as PEG-12 tridecyl ether) used as an emulsifier, solubilizer, and wetting agent in biochemical, formulation, and personal-care applications. It offers good water solubility and emulsion-stabilizing properties, and is also used as a raw material for producing other surfactants and specialty chemicals.
  • Nonionic surfactant for emulsification and solubilization.
  • Good water solubility across formulation types.
  • Low toxicity and mild handling profile.
  • Effective wetting agent for surface treatments.
  • Useful intermediate for producing specialty surfactants.