Learn More
Medchemexpress LLC Polyoxyl 100 stearate | 9004-99-3 | 98.0% | 4686.89 g/mol | C218H436O102 | 100 G
SDP
Catalog No. 5000248361
Encompass_Preferred
Polyoxyl 100 stearate (PEG-100 stearate) is a nonionic polyethylene glycol ester surfactant used as a pharmaceutical excipient. It is commonly employed in formulations as an oil-in-water emulsifier and solubilizer, and is suitable for topical and solid dosage form bases. Supplied as a solid with typical purity around 98% (CAS 9004-99-3) for research use.
  • Acts as o/w emulsifier for stable emulsions.
  • Enhances solubility of lipophilic active ingredients.
  • Suitable for ointment and suppository bases.
  • Solid form simplifies handling and incorporation into formulations.
  • High purity supports formulation consistency and reproducibility.
Catalog No. 50-002-48361 Supplier Medchemexpress LLC Supplier No. HY101530C100G
Polyoxyl 100 stearate (PEG-100 stearate) is a nonionic polyethylene glycol ester surfactant used as a pharmaceutical excipient. It is commonly employed in formulations as an oil-in-water emulsifier and solubilizer, and is suitable for topical and solid dosage form bases. Supplied as a solid with typical purity around 98% (CAS 9004-99-3) for research use.
  • Acts as o/w emulsifier for stable emulsions.
  • Enhances solubility of lipophilic active ingredients.
  • Suitable for ointment and suppository bases.
  • Solid form simplifies handling and incorporation into formulations.
  • High purity supports formulation consistency and reproducibility.