Learn More
Medchemexpress LLC Polyoxyl 100 stearate | 9004-99-3 | 98.0% | 4686.89 g/mol | C218H436O102 | 25 G
SDP
Catalog No. 5000248363
Encompass_Preferred
Polyoxyl 100 stearate (PEG-100 stearate, CAS 9004-99-3) is a high-molecular-weight, nonionic surfactant and pharmaceutical excipient used in laboratory and formulation work. It functions as an oil-in-water emulsifier and solubilizer and serves as a base for ointments and suppositories. Recommended storage is 4°C in sealed containers away from moisture.
  • Acts as a nonionic oil-in-water emulsifier for stable formulations.
  • Enhances solubility of lipophilic compounds.
  • Suitable as an ointment and suppository base for topical applications.
  • High molecular weight provides formulation stability and viscosity control.
  • Storage conditions: sealed at 4°C to maintain integrity.
Catalog No. 50-002-48363 Supplier Medchemexpress LLC Supplier No. HY101530C25G
Polyoxyl 100 stearate (PEG-100 stearate, CAS 9004-99-3) is a high-molecular-weight, nonionic surfactant and pharmaceutical excipient used in laboratory and formulation work. It functions as an oil-in-water emulsifier and solubilizer and serves as a base for ointments and suppositories. Recommended storage is 4°C in sealed containers away from moisture.
  • Acts as a nonionic oil-in-water emulsifier for stable formulations.
  • Enhances solubility of lipophilic compounds.
  • Suitable as an ointment and suppository base for topical applications.
  • High molecular weight provides formulation stability and viscosity control.
  • Storage conditions: sealed at 4°C to maintain integrity.