Learn More
Medchemexpress LLC Polyoxyl 20 Cetostearyl Ether | 68439-49-6 | 10 G
SDP
Catalog No. 5000376454
Encompass_Preferred
Polyoxyl 20 Cetostearyl Ether serves as a pharmaceutical excipient, functioning as an emulsifier and solubilizer in formulations like emulsions and creams. As an inactive ingredient, it enhances the stability, solubility, and processability of pharmaceutical preparations, and may also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Functions as an emulsifier and solubilizer
  • Enhances stability of pharmaceutical preparations
  • Improves solubility of pharmaceutical preparations
  • Facilitates processability of pharmaceutical preparations
  • May influence ADME processes of co-administered drugs
Catalog No. 50-003-76454 Supplier Medchemexpress LLC Supplier No. HY15463910G
Add to Cart
Add to Cart
Polyoxyl 20 Cetostearyl Ether serves as a pharmaceutical excipient, functioning as an emulsifier and solubilizer in formulations like emulsions and creams. As an inactive ingredient, it enhances the stability, solubility, and processability of pharmaceutical preparations, and may also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Functions as an emulsifier and solubilizer
  • Enhances stability of pharmaceutical preparations
  • Improves solubility of pharmaceutical preparations
  • Facilitates processability of pharmaceutical preparations
  • May influence ADME processes of co-administered drugs