Learn More
Medchemexpress LLC Polyoxyl 40 stearate | 171286-89-8 | 98.0% | 2046.58 g/mol | C98H196O42 | 1 G
SDP
Catalog No. 5000248005
Encompass_Preferred
Polyoxyl 40 stearate is a polyethylene glycol monostearate (PEG-40 stearate) used as a nonionic surfactant and pharmaceutical excipient. Supplied as a white to off-white solid for formulation and analytical use, the material is typically reported with high purity and is used to aid emulsification and solubilization of lipophilic ingredients.
  • Used as a nonionic emulsifier and surfactant.
  • Suitable for pharmaceutical formulation and analytical use.
  • Improves solubility and dispersion of lipophilic compounds.
  • Available in small laboratory pack sizes for research use.
  • High reported purity supports analytical applications.
Catalog No. 50-002-48005 Supplier Medchemexpress LLC Supplier No. HY101530B1G
Polyoxyl 40 stearate is a polyethylene glycol monostearate (PEG-40 stearate) used as a nonionic surfactant and pharmaceutical excipient. Supplied as a white to off-white solid for formulation and analytical use, the material is typically reported with high purity and is used to aid emulsification and solubilization of lipophilic ingredients.
  • Used as a nonionic emulsifier and surfactant.
  • Suitable for pharmaceutical formulation and analytical use.
  • Improves solubility and dispersion of lipophilic compounds.
  • Available in small laboratory pack sizes for research use.
  • High reported purity supports analytical applications.