Learn More
Tisch Scientific POLYP MEM FLTR 10UM 47MM 200PK
SDP
Catalog No. NC2246137
Encompass
Polypropylene Membrane Filters, 10.0 um, 47mm, Nonsterile, 200 per pack, SF14907.
This item is not returnable. View return policy
Catalog No. NC2246137 Supplier Tisch Scientific Supplier No. SF14907
Polypropylene Membrane Filters, 10.0 um, 47mm, Nonsterile, 200 per pack, SF14907.