Learn More
Medchemexpress LLC HY-N0047 5mg Medchemexpress, Polyphyllin I CAS:50773-41-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00475MG
Encompass_Preferred
Medchemexpress, HY-N0047 5mg Polyphyllin I CAS:50773-41-6 Polyphyllin I is a bioactive constituent extracted from Paris polyphylla, has strong anti-tumor activity. Polyphyllin I is an activator of the JNK signaling pathway and is an inhibitor of PDK1/Akt/mTOR signaling. Polyphyllin I induces autophagy, G2/M phase arrest and apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1298