Learn More
Medchemexpress LLC HY-N0816 5mg Medchemexpress, Polyphyllin VI CAS:55916-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08165MG
Encompass_Preferred
Medchemexpress, HY-N0816 5mg Polyphyllin VI CAS:55916-51-3 Polyphyllin VI, an active saponin mainly isolated from traditional medicinal plant Paris polyphylla, possess anti-cancer activities. Polyphyllin VI induces G2/M cell cycle arrest and triggers apoptosis[2]. Polyphyllin VI induces caspase-1-mediated pyroptosis via the induction of ROS/NF-κB/NLRP3/GSDMD signal axis in non-small cell lung cancer[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1407