Learn More
MSC Carlisle Bi-Level Scrub Brush, 10 in, Blue, Polypropylene, 1 ea
Catalog No. 502866471
Encompass
This bi-level scrub brush is designed for efficient cleaning of various surfaces. Its split-shape design easily cleans flat surfaces, under counters, around equipment, and along baseboards. The bristles are nonabsorbent, making them resistant to oils and moisture, and the brush is built for durability.
  • Split shape effectively cleans diverse surfaces and hard-to-reach areas
  • Bristles are resistant to oils and moisture, preventing mold and mildew growth
  • Hi-Lo design offers maximum cleaning efficiency from three different angles
  • Foam-injected block provides enhanced durability for long-term use
  • Features a standard 3/4-5 ACME thread for compatible handles
  • Color-coded for HACCP practices to prevent cross-contamination
Catalog No. 50-286-6471 Supplier MSC Supplier No. 10785483
This bi-level scrub brush is designed for efficient cleaning of various surfaces. Its split-shape design easily cleans flat surfaces, under counters, around equipment, and along baseboards. The bristles are nonabsorbent, making them resistant to oils and moisture, and the brush is built for durability.
  • Split shape effectively cleans diverse surfaces and hard-to-reach areas
  • Bristles are resistant to oils and moisture, preventing mold and mildew growth
  • Hi-Lo design offers maximum cleaning efficiency from three different angles
  • Foam-injected block provides enhanced durability for long-term use
  • Features a standard 3/4-5 ACME thread for compatible handles
  • Color-coded for HACCP practices to prevent cross-contamination