Learn More
Medchemexpress LLC Polyporenic acid C | 465-18-9 | 5 MG
SDP
Catalog No. 5000420671
Encompass_Preferred
Polyporenic acid C is a lanostane-type triterpenoid isolated from Poria cocos. This compound induces apoptosis and exhibits anticancer activity, particularly against non-small cell lung cancer, by affecting various cellular pathways.
  • Induces the cleavage of caspase-8 and caspase-3
  • Causes the cleavage of PARP
  • Reduces the phosphorylation level of Akt (Ser473)
  • Increases the phosphorylation of PTEN and p53 (Ser15)
  • Activates JNK
Catalog No. 50-004-20671 Supplier Medchemexpress LLC Supplier No. HYN29935MG
Add to Cart
Add to Cart
Polyporenic acid C is a lanostane-type triterpenoid isolated from Poria cocos. This compound induces apoptosis and exhibits anticancer activity, particularly against non-small cell lung cancer, by affecting various cellular pathways.
  • Induces the cleavage of caspase-8 and caspase-3
  • Causes the cleavage of PARP
  • Reduces the phosphorylation level of Akt (Ser473)
  • Increases the phosphorylation of PTEN and p53 (Ser15)
  • Activates JNK