Learn More
Medchemexpress LLC HY-N7693 1mg Medchemexpress, Polyporusterone B CAS:141360-89-6 Purity:>98%
Supplier: Medchemexpress LLC HYN76931MG
Medchemexpress, HY-N7693 1mg Polyporusterone B CAS:141360-89-6 Polyporusterone B is a triterpene carboxylic acid isolated from Polyporus umbellatus Fries. Polyporusterone B has inhibitory effect on free radical-induced lysis of red blood cells (hemolysis). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.