Learn More
SCIPRO INC POLYPR.SAMPLE VIALS 1000/CS
Catalog No. NC9010038
Encompass
Polypropylene Sample Vial, The sample vials fit with most fraction collectors used in microdialysis, The vials can be used for collecting sample with or without the re-sealing caps, 300uL, 1000/PK
Catalog No. NC9010038 Supplier SCIPRO INC Supplier No. 4001048
Polypropylene Sample Vial, The sample vials fit with most fraction collectors used in microdialysis, The vials can be used for collecting sample with or without the re-sealing caps, 300uL, 1000/PK