Learn More
Business Communication Solutions 2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label, Permanent Adhesive, Floodcoat UV Varnish, 510 Labels/Roll, 9 Rolls/Case, 0.75" Core, 2.375" OD.
Catalog No. NC1593771
Encompass_Preferred
2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label, Permanent Adhesive, IPA Resistant Varnish, 510 Labels/Roll, 9 Rolls/Case, .75" Core, 2.8715" OD. Compatible with Zebra Model ZQ620 and QLn320 Thermal Barcode Printers. One year expiration.
Catalog No. NC1593771 Supplier Business Communication Solutions Supplier No. 225125PPDUV/DEL
2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label, Permanent Adhesive, IPA Resistant Varnish, 510 Labels/Roll, 9 Rolls/Case, .75" Core, 2.8715" OD. Compatible with Zebra Model ZQ620 and QLn320 Thermal Barcode Printers. One year expiration.