Learn More
MSC Vikan Lid for Bucket, 5 Gallon, Polypropylene, Blue
Catalog No. 502835771
Encompass
This lid is designed for a 5 gallon bucket, preventing spills and protecting contents from contaminants during storage or transport. Constructed from ultra-durable polypropylene, it features a special stacking rim and a clip system for a secure fit, allowing compatible buckets to be stacked for space-saving storage. It is also ideal for color-coded organization.
  • Prevents spills and protects contents from contaminants.
  • Features a stacking rim for space-saving storage.
  • Securely snaps onto compatible buckets.
  • Constructed from ultra-durable polypropylene.
  • Ideal for color-coded organization.
  • Complies with food contact material regulations (EC 1935/2004, FDA CFR 21, UK 2019 No. 704).
  • Dishwasher safe.
Catalog No. 50-283-5771 Supplier MSC Supplier No. 54799648
This lid is designed for a 5 gallon bucket, preventing spills and protecting contents from contaminants during storage or transport. Constructed from ultra-durable polypropylene, it features a special stacking rim and a clip system for a secure fit, allowing compatible buckets to be stacked for space-saving storage. It is also ideal for color-coded organization.
  • Prevents spills and protects contents from contaminants.
  • Features a stacking rim for space-saving storage.
  • Securely snaps onto compatible buckets.
  • Constructed from ultra-durable polypropylene.
  • Ideal for color-coded organization.
  • Complies with food contact material regulations (EC 1935/2004, FDA CFR 21, UK 2019 No. 704).
  • Dishwasher safe.