Learn More
MSC Vikan Pipe Cleaning Brush, Ø50 mm, Hard, Yellow
Catalog No. 502837571
Encompass
This versatile cleaning brush is designed to thoroughly clean the inside of pipes, tubes, and other hard-to-reach spaces, making it ideal for various applications including food processing equipment. Featuring a durable polypropylene shaft and stiff polyester bristles, it effectively removes stubborn dirt, grime, and residue.
  • Cleans the inside of pipes and tubes effectively.
  • Features a durable polypropylene shaft and stiff polyester bristles.
  • Efficiently removes stubborn dirt, grime, and residue.
  • Yellow color supports HACCP color-coded systems.
  • Compatible with any cleaning handle for versatile use.
  • Certified for food contact by EU 10/2011 and FDA CFR 21.
  • Dishwasher and autoclave safe for thorough sanitization.
  • Ideal for cleaning food processing machinery like meat mincers.
  • Designed for repeated, long-term use.
Catalog No. 50-283-7571 Supplier MSC Supplier No. 47173307
This versatile cleaning brush is designed to thoroughly clean the inside of pipes, tubes, and other hard-to-reach spaces, making it ideal for various applications including food processing equipment. Featuring a durable polypropylene shaft and stiff polyester bristles, it effectively removes stubborn dirt, grime, and residue.
  • Cleans the inside of pipes and tubes effectively.
  • Features a durable polypropylene shaft and stiff polyester bristles.
  • Efficiently removes stubborn dirt, grime, and residue.
  • Yellow color supports HACCP color-coded systems.
  • Compatible with any cleaning handle for versatile use.
  • Certified for food contact by EU 10/2011 and FDA CFR 21.
  • Dishwasher and autoclave safe for thorough sanitization.
  • Ideal for cleaning food processing machinery like meat mincers.
  • Designed for repeated, long-term use.