Learn More
Andwin Scientific Polypropylene, Large Screw Cap 15ml Centrifuge, 500/pack
SDP
Catalog No. 50727623
Encompass_Preferred
Constructed from polypropylene, this Large Screw Cap 15ml Centrifuge Tube withstands 1800RCF with glass-like clarity and chemical resistance up to 6400 RCF. It is available in 500 units per pack.
Catalog No. 50-727-623 Supplier Andwin Scientific Supplier No. 6476420
Constructed from polypropylene, this Large Screw Cap 15ml Centrifuge Tube withstands 1800RCF with glass-like clarity and chemical resistance up to 6400 RCF. It is available in 500 units per pack.