Learn More
Grainger Sterilizing Tray: Polypropylene, 5 1/8 in Ht, 10 1/2 in Wd, 19 in Lg
Supplier:  Grainger 46C854
Encompass_Preferred
This molded plastic tray has rounded corners and smooth, tapered sides for stacking. It's ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks. The tray is steam autoclavable at 121°C.
Catalog No. NC1789257