Learn More
Cole-Parmer Cole-Parmer Polypropylene utility tray, 18" x 18" x 4"
Supplier:  Cole-Parmer 0671075
Encompass_Preferred
This utility tray is 3/16 inch thick and made of polypropylene. It also is autoclavable.
Catalog No. NC1478990