Learn More
Polylc Inc POLYPROPYL A 100X4.6MM 2UM
SDP
Catalog No. NC1700022
Encompass
PolyPROPYL A column, 100x4.6mm (2μm;1500Å)
Catalog No. NC1700022 Supplier Polylc Inc Supplier No. 104PR0215
PolyPROPYL A column, 100x4.6mm (2μm;1500Å)