Learn More
Andwin Scientific POLYPROPYLENE 12CH BASIN 20PK
SDP
Catalog No. NC1677308
Encompass_Preferred
Reagent Bason^ Argos sterile, polypropylene 12 $246.3200 $141.0000 $141.0000channel basin is ideal for use with a multichannelpipettor for performing dilutions. Each numericallymarked channel holds 5 ml of sample. V shapedbottom for complete sample recovery. With Fittedlid, 20/pk
Catalog No. NC1677308 Supplier Andwin Scientific Supplier No. 89029706
Reagent Bason^ Argos sterile, polypropylene 12 $246.3200 $141.0000 $141.0000channel basin is ideal for use with a multichannelpipettor for performing dilutions. Each numericallymarked channel holds 5 ml of sample. V shapedbottom for complete sample recovery. With Fittedlid, 20/pk