Learn More
Revvity Health Sciences Inc Microplate 384-well, (U)-bottom, polypropylene, low volume, 35 µL, case of 50
Catalog No. 502106051
Encompass
Microplate 384-well, (U)-bottom, polypropylene, low volume, 35 µL, case of 50
Catalog No. 50-210-6051 Supplier Revvity Health Sciences Inc Supplier No. 6008890
Microplate 384-well, (U)-bottom, polypropylene, low volume, 35 µL, case of 50