Learn More
USA Scientific Inc 50-place PP Box
Catalog No. NC9051372
Encompass
Polypropylene 50-place box, natural,  5 2/3 in L x 3 in W x 2 in H, 5/pack
Catalog No. NC9051372 Supplier USA Scientific Inc Supplier No. 23500500
Polypropylene 50-place box, natural,  5 2/3 in L x 3 in W x 2 in H, 5/pack