Learn More
Cole-Parmer Hollow Polypropylene (PP) Ball Bath Covers, 20 mm; 500/Pk
Supplier:  Cole-Parmer 0682102
Encompass
Hollow Polypropylene (PP) Ball Bath Covers, 20 mm; 500/Pk, Reduce vapor and heat loss by 75% with one layer of Bath balls, Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.
Catalog No. NC1219054