Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Polypropylene Beard Restraints

Catalog No. 19472785 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-472-785 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-472-785 Supplier International Enviroguard™ Supplier No. S112NW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beard restraints provide economical protection against dirt, grime, and particulates in non-hazardous environments.

  • Polypropylene
  • Provides particulate barrier
  • Helps prevent facial hair from contaminating a product or work environment
  • Low-linting, lightweight
  • Restrains facial hair and offers sufficient ventilation
  • Protects against dirt, grime, and particulates in non-hazardous environments
  • Latex-free elastic
  • Helps to prevent cross-contamination of food items or sensitive items like microelectronics
  • Possible applications include food processing, food service, cleanrooms, controlled environments, pharmaceuticals, environmental cleanup

Specifications

Type Cleanroom Beard Cover
Size Universal
Material Polypropylene
Color White
Disposable Single-use
Latex Free Yes
For Use With (Application) Food Processing, Food Service, Cleanrooms, Pharmaceuticals, Environmental Cleanup
Length (English) 7 in.
Length (Metric) 17.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.