Learn More
Waters Corp Waters Polypropylene Cap Mat, Square Well, 2 mL, 96-well Plate
Catalog No. 50781143 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polypropylene cap mat serves as a sealing option for 96-well plates with a 2 mL well volume. It is designed to cover all 96 wells of sample or elution plates, ensuring short- or long-term preservation of chemicals or molecules. This helps prevent contamination and evaporation of well contents, contributing to pure analytes and consistent, accurate findings.
  • Made from polypropylene for broad solvent resistance and good sealing properties.
  • Designed to cover all 96 wells for sample or elution plates.
  • Helps prevent contamination and evaporation of well contents.
  • Compatible with 96-well plates and square well 96-well plates.
  • Suitable for use with 700 μL glass inserts.
  • Approved for use with certain ACQUITY UPLC configurations.
  • Packaged as 50 pieces per pack.
Catalog No. 50-781-143 Supplier Waters Corp Supplier No. 186002484
This polypropylene cap mat serves as a sealing option for 96-well plates with a 2 mL well volume. It is designed to cover all 96 wells of sample or elution plates, ensuring short- or long-term preservation of chemicals or molecules. This helps prevent contamination and evaporation of well contents, contributing to pure analytes and consistent, accurate findings.
  • Made from polypropylene for broad solvent resistance and good sealing properties.
  • Designed to cover all 96 wells for sample or elution plates.
  • Helps prevent contamination and evaporation of well contents.
  • Compatible with 96-well plates and square well 96-well plates.
  • Suitable for use with 700 μL glass inserts.
  • Approved for use with certain ACQUITY UPLC configurations.
  • Packaged as 50 pieces per pack.