Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Polypropylene, Carboy with 83 mm Cap
GREENER_CHOICE

Catalog No. 19830432 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
5.3 gal.
Capacity (Metric):
20 L
Dimensions (L x W x H):
345 x 206 x 615 mm
Closure Size:
83 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Dimensions (L x W x H) Closure Size
19-830-432 5.3 gal. 20 L 345 x 206 x 615 mm 83 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-830-432 Supplier Justrite™ Supplier No. 12931
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials

Specifications

Accuracy ±5%
Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 345 x 206 x 615 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 24.2 in.
Length (English) 13.6 in.
Material Polypropylene
Product Type Carboy
Shape Rectangular
Width (English) 8.1 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.