Learn More
Sigma Aldrich Fine Chemicals Biosciences Polypropylene carboy size1EA
Catalog No. 502900624
Encompass
All polypropylene construction is autoclavable 121C (250f) and chemical resistant. When autoclaving do not install cap.83 mm cap seals with a silicone gasketOne large handle for transport or pouring
Catalog No. 50-290-0624 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6755631EA
All polypropylene construction is autoclavable 121C (250f) and chemical resistant. When autoclaving do not install cap.83 mm cap seals with a silicone gasketOne large handle for transport or pouring