Learn More
Sigma Aldrich Fine Chemicals Biosciences Polypropylene carboy with1EA
Catalog No. 502901638
Encompass
Heavy-duty carboys have a molded-in threaded bung that receives the spigot. All polypropylene construction is autoclavable 121C (250f) and chemical resistant. When autoclaving do not install cap.Single-handed spigot control for simple dispensing83 mm cap seals with a silicone gasketTwo large handles for transport or pouring
Catalog No. 50-290-1638 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6755391EA
Heavy-duty carboys have a molded-in threaded bung that receives the spigot. All polypropylene construction is autoclavable 121C (250f) and chemical resistant. When autoclaving do not install cap.Single-handed spigot control for simple dispensing83 mm cap seals with a silicone gasketTwo large handles for transport or pouring