Learn More
Biosurface Technologies Corp Polypropylene Disc Coupon, 0.5 inch diameter for use with CDC, IBR, and RDR Biofilm Reactors
Catalog No. NC0548988
Encompass
This 0.5 inch (12.7mm) diameter coupon is manufactured from polypropylene, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 8 per supplier requirements.
Catalog No. NC0548988 Supplier Biosurface Technologies Corp Supplier No. RD128PP
This 0.5 inch (12.7mm) diameter coupon is manufactured from polypropylene, and is designed for biofilm or biofilm-related studies in a laboratory environment using the CDC Biofilm Reactor, Bio-inLine Biofilm Reactor, or the Rotating Disk Biofilm Reactor. This item has a minimum quantity of 8 per supplier requirements.