Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Polypropylene Coveralls with Elastic Wrists and Open Ankles

Catalog No. 17184239 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-239 2X-Large
17-184-235 Large
17-184-233 X-Large
17-184-238 3X-Large
17-184-236 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-184-239 Supplier International Enviroguard™ Supplier No. 20122XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disposable coveralls provide low-cost protection against dust, dirt, and grime.

  • Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
  • Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
  • Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
  • Elastic wrists, open ankles
  • Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
  • Stay cool, comfortable, and protected from head-to-toe
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more

Specifications

Product Type Light-duty Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Dust
Collar Type Fold-down
Latex Free Yes
Model 2012
No. of Pockets None
For Use With (Application) Food Packaging, Food Processing, Warehousing, Laboratories
Length (English) 40.5 in.
Length (English) Sleeve 36.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.