Learn More
Ted Pella Inc Static-Free Plastic Cups, 30 mL, 100/pk
SDP
Supplier:  Ted Pella Inc 12901
Encompass
1 fl. oz. volume cups (30ml) which are ideal for preparing water for glass or diamond knife troughs. Causes distilled water to be static-free. Serial sections will not move around the boat. These plastic cups are great for working with ribbons of sections, and manipulating electron microscopy grids. Graduated by side markings in CC, ML, OZ. and Drams.
Catalog No. NC9425468