Learn More
BUCHI Polyethylene Frits Set for Multivapor™
SDP
Catalog No. 05405916 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Multivapor™
Catalog No. For Use With (Equipment)
05-405-916 Multivapor™
1 options
Catalog No. 05-405-916 Supplier BUCHI Supplier No. 044856
Add to Cart
Add to Cart
Polyethylene frits to protect against cross-contamination of powders or solvent bumping.
  • Chemically resistant polyethylene
  • Helps protect against cross-contamination and sample loss
  • Designed for Multivapor™
  • Frit diameter: 10 mm
  • 64 Pk.
Specifications
Material Polyethylene
For Use With (Application) Protection Against Cross-contamination and Solvent Bumping
For Use With (Equipment) Multivapor™
Diameter (Metric) 10 mm